Gene Transcripts

Gene Symbol
HSD17B12
Entrez Gene ID
51144
Gene Name
hydroxysteroid 17-beta dehydrogenase 12
Gene Aliases
KAR, SDR12C1
Chromosome Location
Chr. 11: 43680593 - 43856619 on Build GRCh38
UniGene ID
Hs.132513
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016142.2 NP_057226.1 8 827
XM_011520156.1 XP_011518458.1 8 821
XM_017017881.1 XP_016873370.1 8 507
GenBank mRNA AF078850.1 - 8 665
AK027882.1 - 720
AK074952.1 - 8 679
AK075011.1 - 8 712
AK075216.1 - 8 688
AK092938.1 - 1004
AK222881.1 - 8 649
AK292625.1 - 8 712
AK301663.1 - 9 618
BC012043.1 - 8 650 3622879

View Full Product Details