Gene Transcripts

Gene Symbol
BORCS6
Entrez Gene ID
54785
Gene Name
BLOC-1 related complex subunit 6
Gene Aliases
C17orf59, PRO2472
Chromosome Location
Chr. 17: 8188333 - 8190246 on Build GRCh38
UniGene ID
Hs.129563
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017622.2 NP_060092.2 1 1146
GenBank mRNA AI174760.1 - 1 165
AK000021.1 - 1 972
AK222506.1 - 1 972
BC009261.2 - 1 929 3928189
BC018880.2 - 1 1028 3615792
BC070272.1 - 1 1043 4672989

View Full Product Details