Gene Transcripts

Gene Symbol
TTC19
Entrez Gene ID
54902
Gene Name
tetratricopeptide repeat domain 19
Gene Aliases
2010204O13Rik, MC3DN2
Chromosome Location
Chr. 17: 15999380 - 16045428 on Build GRCh38
UniGene ID
Hs.462316
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271420.1 NP_001258349.1 8 1402
NM_017775.3 NP_060245.3 8 1265
XM_017024801.1 XP_016880290.1 8 1178
XM_017024802.1 XP_016880291.1 8 1178
GenBank mRNA AK000350.1 - 6 481
AK025958.1 - 2 151
AK055780.1 - 8 958
AK056878.1 - 8 1177
AK094819.1 - 1 1459
AK297783.1 - 8 821
AL832412.1 - 1 519
BC011698.2 - 8 804
BC073796.1 - 8 808
BC105128.1 - 8 1265 8144131
BC112107.1 - 8 1265 8327575

View Full Product Details