Gene Transcripts

Gene Symbol
TTC19
Entrez Gene ID
54902
Gene Name
tetratricopeptide repeat domain 19
Gene Aliases
2010204O13Rik, MC3DN2
Chromosome Location
Chr. 17: 15999380 - 16045428 on Build GRCh38
UniGene ID
Hs.462316
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271420.1 NP_001258349.1 9 1528
NM_017775.3 NP_060245.3 9 1391
XM_017024801.1 XP_016880290.1 9 1304
XM_017024802.1 XP_016880291.1 9 1304
GenBank mRNA AK000350.1 - 7 607
AK025958.1 - 3 277
AK055780.1 - 9 1084
AK056878.1 - 9 1303
AK094819.1 - 2 1585
AK297783.1 - 9 947
AL832412.1 - 2 645
BC011698.2 - 9 930
BC073796.1 - 9 934
BC105128.1 - 9 1391 8144131
BC112107.1 - 9 1391 8327575

View Full Product Details