Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017820.4 | NP_060290.3 | 20 | 2392 | |
XM_011518810.1 | XP_011517112.1 | 22 | 2857 | ||
XM_011518814.1 | XP_011517116.1 | 14 | 1807 | ||
XM_011518815.1 | XP_011517117.1 | 11 | 1491 | ||
XM_017014861.1 | XP_016870350.1 | 23 | 3001 | ||
XM_017014862.1 | XP_016870351.1 | 21 | 2742 | ||
XM_017014863.1 | XP_016870352.1 | 21 | 2700 | ||
XM_017014864.1 | XP_016870353.1 | 20 | 3062 | ||
XM_017014865.1 | XP_016870354.1 | 20 | 2709 | ||
XM_017014866.1 | XP_016870355.1 | 19 | 2365 | ||
XM_017014867.1 | XP_016870356.1 | 19 | 2406 | ||
GenBank mRNA | AB061798.1 | - | 14 | 1741 | |
AK055004.1 | - | 10 | 1524 | ||
AK094438.1 | - | 20 | 2292 | ||
BU947354.1 | - | 2 | 114 |