Gene Transcripts
- Gene Symbol
- LRRC1
- Entrez Gene ID
- 55227
- Gene Name
- leucine rich repeat containing 1
- Gene Aliases
- LANO, dJ523E19.1
- Chromosome Location
- Chr. 6: 53794497 - 53924125 on Build GRCh38
- UniGene ID
- Hs.606493
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018214.4 |
NP_060684.4
|
5, 6 |
773 |
|
XM_011514726.2 |
XP_011513028.1
|
6, 7 |
1137 |
|
XM_011514727.2 |
XP_011513029.1
|
4, 5 |
563 |
|
XM_017010997.1 |
XP_016866486.1
|
5, 6 |
1065 |
|
GenBank mRNA |
AF332199.1 |
- |
5, 6 |
720 |
|
AF359380.1 |
- |
5, 6 |
496 |
|
AK001637.1 |
- |
5, 6 |
477 |
|
AK021896.1 |
- |
6, 7 |
1137 |
|
AK292248.1 |
- |
5, 6 |
618 |
|
BC003193.1 |
- |
5, 6 |
773 |
3504090 |
View Full Product Details