Gene Transcripts

Gene Symbol
SMU1
Entrez Gene ID
55234
Gene Name
DNA replication regulator and spliceosomal factor
Gene Aliases
BWD, SMU-1, fSAP57
Chromosome Location
Chr. 9: 33041852 - 33076716 on Build GRCh38
UniGene ID
Hs.655351
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018225.2 NP_060695.2 10 1298
XM_005251503.4 XP_005251560.1 9 1134
GenBank mRNA AK001667.1 - 10 1228
AK022032.1 - 2 102
AK225559.1 - 10 1253
AK225587.1 - 10 1247
AK304767.1 - 8 885
BC002876.2 - 10 1244 3940669
BC110654.1 - 10 1338 6168993
EF011613.1 - 10 1244

View Full Product Details