Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001184977.1 | NP_001171906.1 | 2 | 227 | |
NM_018244.4 | NP_060714.3 | 2 | 227 | ||
NM_199487.2 | NP_955781.2 | 2 | 227 | ||
XM_011528877.1 | XP_011527179.1 | 2 | 132 | ||
XM_011528878.1 | XP_011527180.1 | 2 | 132 | ||
XM_011528879.1 | XP_011527181.1 | 2 | 78 | ||
XM_011528880.1 | XP_011527182.1 | 2 | 136 | ||
XM_011528881.2 | XP_011527183.1 | 2 | 254 | ||
XM_011528882.1 | XP_011527184.1 | 2 | 85 | ||
XM_011528883.1 | XP_011527185.1 | 2 | 107 | ||
XM_011528884.1 | XP_011527186.1 | 2 | 58 | ||
GenBank mRNA | AK001712.1 | - | 2 | 58 | |
AK022650.1 | - | 2 | 58 | ||
AK022742.1 | - | 3 | 181 | ||
AK225281.1 | - | 2 | 67 | ||
AK225557.1 | - | 2 | 58 | ||
AK293516.1 | - | 2 | 58 | ||
AK300130.1 | - | 2 | 58 | ||
AK302133.1 | - | 2 | 58 | ||
AK308819.1 | - | 2 | 58 | ||
AU280736.1 | - | 2 | 58 | ||
AY050659.1 | - | 2 | 227 | ||
AY050660.1 | - | 1 | 30 | ||
BC105791.1 | - | 2 | 62 | 6458054 | |
BC119013.1 | - | 2 | 93 | 40085306 | |
BC122862.1 | - | 2 | 93 | 40085305 | |
BM919736.1 | - | 2 | 80 |