Gene Transcripts

Gene Symbol
KDM4D
Entrez Gene ID
55693
Gene Name
lysine demethylase 4D
Gene Aliases
JMJD2D
Chromosome Location
Chr. 11: 94973681 - 94999514 on Build GRCh38
UniGene ID
Hs.503598
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018039.2 NP_060509.2 3 2064
GenBank mRNA AK056162.1 - 1 1476
AL137545.1 - 1 52
BC074739.2 - 1 727

View Full Product Details