Gene Transcripts
- Gene Symbol
- NDC1
- Entrez Gene ID
- 55706
- Gene Name
- NDC1 transmembrane nucleoporin
- Gene Aliases
- NET3, TMEM48
- Chromosome Location
- Chr. 1: 53765460 - 53838552 on Build GRCh38
- UniGene ID
- Hs.476525
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001168551.1 |
NP_001162023.1
|
13, 14 |
1743 |
|
NM_018087.4 |
NP_060557.3
|
13, 14 |
1863 |
|
NR_033142.1 |
|
12, 13 |
1742 |
|
XM_011541766.2 |
XP_011540068.1
|
13, 14 |
1705 |
|
GenBank mRNA |
AK001269.1 |
- |
13, 14 |
1746 |
|
AK022618.1 |
- |
1, 2 |
19 |
|
AK091439.1 |
- |
7, 8 |
549 |
|
AK295000.1 |
- |
13, 14 |
1527 |
|
AK302910.1 |
- |
11, 12 |
1321 |
|
AL354613.1 |
- |
13, 14 |
1743 |
|
BC003082.2 |
- |
13, 14 |
1813 |
3509727 |
BX648031.1 |
- |
12, 13 |
1473 |
|
DQ141696.1 |
- |
13, 14 |
1572 |
|
View Full Product Details