Gene Transcripts

Gene Symbol
SUSD2
Entrez Gene ID
56241
Gene Name
sushi domain containing 2
Gene Aliases
BK65A6.2
Chromosome Location
Chr. 22: 24181476 - 24189106 on Build GRCh38
UniGene ID
Hs.131819
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019601.3 NP_062547.1 12 2106
GenBank mRNA AK026431.1 - 12 2124
AK126105.1 - 8 3861
BC033107.1 - 12 2106 5457186

View Full Product Details