Gene Transcripts
- Gene Symbol
- CCDC180
- Entrez Gene ID
- 100499483
- Gene Name
- coiled-coil domain containing 180
- Gene Aliases
- BDAG1, C9orf174, KIAA1529
- Chromosome Location
- Chr. 9: 97307628 - 97377295 on Build GRCh38
- UniGene ID
- Hs.435629
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020893.2 |
NP_065944.2
|
25 |
3563 |
|
GenBank mRNA |
CR749320.1 |
- |
25 |
3505 |
|
- Gene Symbol
- LOC100499484-C9ORF174
- Entrez Gene ID
- 57653
- Gene Name
- LOC100499484-C9orf174 readthrough
- Gene Aliases
- Chromosome Location
- Chr. 9: 97238426 - 97377295 on Build GRCh38
- UniGene ID
- Hs.435629
- Species
- Homo sapiens
View Full Product Details