Gene Transcripts

Gene Symbol
IGDCC4
Entrez Gene ID
57722
Gene Name
immunoglobulin superfamily DCC subclass member 4
Gene Aliases
DDM36, NOPE
Chromosome Location
Chr. 15: 65381480 - 65423072 on Build GRCh38
UniGene ID
Hs.458607
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020962.2 NP_066013.1 10 1998
XM_011521845.2 XP_011520147.1 10 2178
XM_011521846.2 XP_011520148.1 10 1802
XM_011521847.1 XP_011520149.1 10 1862
XM_017022448.1 XP_016877937.1 10 2180
XM_017022449.1 XP_016877938.1 10 2186
GenBank mRNA AB046848.1 - 4 1058
AB052622.1 - 10 1998

View Full Product Details