Gene Transcripts

Gene Symbol
FKBPL
Entrez Gene ID
63943
Gene Name
FK506 binding protein like
Gene Aliases
DIR1, FKBP4, NG7, WISP39
Chromosome Location
Chr. 6: 32128707 - 32130290 on Build GRCh38
UniGene ID
Hs.520042
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022110.3 NP_071393.2 2 283
GenBank mRNA AK027119.1 - 2 281
AK291418.1 - 2 278
BC004168.2 - 2 268 2820874
BC011966.1 - 2 283 3857383
CR457374.1 - 1 13
KP861813.1 - 2 269

View Full Product Details