Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022166.3 | NP_071449.1 | 6 | 1414 | |
XM_017023539.1 | XP_016879028.1 | 6 | 1467 | ||
XM_017023540.1 | XP_016879029.1 | 6 | 1467 | ||
GenBank mRNA | AJ277441.1 | - | 4 | 933 | |
AJ539163.1 | - | 6 | 1414 |