Gene Transcripts

Gene Symbol
MMS19
Entrez Gene ID
64210
Gene Name
MMS19 homolog, cytosolic iron-sulfur assembly component
Gene Aliases
MET18, MMS19L, hMMS19
Chromosome Location
Chr. 10: 97458324 - 97498794 on Build GRCh38
UniGene ID
Hs.500721
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289403.1 NP_001276332.1 17 1714
NM_001289404.1 NP_001276333.1 17 1779
NM_001289405.1 NP_001276334.1 19 1725
NM_022362.4 NP_071757.4 18 1843
XM_005270035.2 XP_005270092.1 17 1600
XM_005270041.1 XP_005270098.1 16 1488
XM_006717944.3 XP_006718007.2 18 1634
XM_006717945.2 XP_006718008.1 16 2176
XM_011540062.1 XP_011538364.1 17 1446
XM_011540063.2 XP_011538365.1 16 2174
XM_017016515.1 XP_016872004.1 19 1749
XM_017016516.1 XP_016872005.1 19 1746
XM_017016517.1 XP_016872006.1 18 1620
XM_017016518.1 XP_016872007.1 17 1505
XM_017016519.1 XP_016872008.1 17 1455
XM_017016520.1 XP_016872009.1 16 1343
XM_017016521.1 XP_016872010.1 16 1340
XM_017016522.1 XP_016872011.1 18 1741
XM_017016523.1 XP_016872012.1 17 1625
XM_017016524.1 XP_016872013.1 17 1597
XM_017016525.1 XP_016872014.1 16 1367
XM_017016526.1 XP_016872015.1 17 1603
XM_017016527.1 XP_016872016.1 17 2044
XM_017016528.1 XP_016872017.1 15 2015
XM_017016529.1 XP_016872018.1 17 2244
XM_017016530.1 XP_016872019.1 18 1763
XM_017016531.1 XP_016872020.1 16 1461
XM_017016532.1 XP_016872021.1 15 1238
XM_017016533.1 XP_016872022.1 15 1315
XM_017016534.1 XP_016872023.1 16 2171
XM_017016535.1 XP_016872024.1 15 2032
GenBank mRNA AF007151.1 - 4 306
AF319947.1 - 19 1691
AF357881.1 - 18 1642
AJ306408.1 - 18 1625
AK025496.1 - 5 342
AK027710.1 - 15 1183
AK056244.1 - 7 650
AK056581.1 - 13 1101
AK225312.1 - 18 1628
AK298995.1 - 18 1681
AK304287.1 - 17 1508
AK314374.1 - 18 1641
BC002692.1 - 7 605
BC006575.2 - 10 937
BC080532.1 - 17 1558 4889876
BC117129.1 - 18 1641 40125680

View Full Product Details