Gene Transcripts
- Gene Symbol
- EBF2
- Entrez Gene ID
- 64641
- Gene Name
- early B-cell factor 2
- Gene Aliases
- COE2, EBF-2, O/E-3, OE-3
- Chromosome Location
- Chr. 8: 25841730 - 26045124 on Build GRCh38
- UniGene ID
- Hs.491292
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022659.3 |
NP_073150.2
|
13, 14 |
1605 |
|
GenBank mRNA |
AK021562.1 |
- |
6, 7 |
644 |
|
AK304372.1 |
- |
12, 13 |
1253 |
|
AY700779.1 |
- |
5, 6 |
533 |
|
BC069665.1 |
- |
6, 7 |
622 |
|
BC069726.1 |
- |
6, 7 |
625 |
|
BC069747.1 |
- |
6, 7 |
624 |
|
BC069768.1 |
- |
6, 7 |
625 |
|
BC074794.2 |
- |
5, 6 |
561 |
|
BC113478.1 |
- |
5, 6 |
589 |
|
BC113504.1 |
- |
5, 6 |
589 |
|
View Full Product Details