Gene Transcripts

Gene Symbol
IQCH
Entrez Gene ID
64799
Gene Name
IQ motif containing H
Gene Aliases
NYDSP5
Chromosome Location
Chr. 15: 67254772 - 67501804 on Build GRCh38
UniGene ID
Hs.657894
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031715.2 NP_001026885.1 12 1566
NM_001284347.1 NP_001271276.1 8 1103
NM_001284348.1 NP_001271277.1 10 1338
NM_001322470.1 NP_001309399.1 9 1232
NM_001322471.1 NP_001309400.1 8 1103
NM_001322472.1 NP_001309401.1 8 1103
NM_001322473.1 NP_001309402.1 9 1232
NM_001322474.1 NP_001309403.1 9 1232
NM_001322475.1 NP_001309404.1 10 1350
XM_011521907.2 XP_011520209.1 13 1707
XM_011521908.2 XP_011520210.1 12 1566
XM_011521911.2 XP_011520213.1 11 1494
XM_011521912.2 XP_011520214.1 13 1707
XM_011521913.2 XP_011520215.1 13 1708
XM_011521914.2 XP_011520216.1 10 1226
XM_011521915.2 XP_011520217.1 13 1707
XM_011521916.2 XP_011520218.1 9 1145
XM_011521917.2 XP_011520219.1 11 1414
XM_011521918.2 XP_011520220.1 9 1200
XM_017022491.1 XP_016877980.1 13 1707
XM_017022492.1 XP_016877981.1 13 1707
XM_017022493.1 XP_016877982.1 12 1668
XM_017022494.1 XP_016877983.1 13 1707
XM_017022495.1 XP_016877984.1 13 1707
XM_017022496.1 XP_016877985.1 13 1707
XM_017022497.1 XP_016877986.1 13 1707
XM_017022498.1 XP_016877987.1 13 1707
XM_017022499.1 XP_016877988.1 13 1707
XM_017022500.1 XP_016877989.1 10 1373
XM_017022501.1 XP_016877990.1 9 1200
XM_017022502.1 XP_016877991.1 10 1361
XM_017022503.1 XP_016877992.1 8 1126
XM_017022504.1 XP_016877993.1 8 1126
GenBank mRNA AK022538.1 - 10 1288
AK292478.1 - 12 1518
AY014282.2 - 12 1537
BC039292.1 - 8 1053 5270043
BC048964.1 - 12 1516 5297681

View Full Product Details