Gene Transcripts
- Gene Symbol
- CYP4F12
- Entrez Gene ID
- 66002
- Gene Name
- cytochrome P450 family 4 subfamily F member 12
- Gene Aliases
- CYPIVF12, F22329_1
- Chromosome Location
- Chr. 19: 15673018 - 15698819 on Build GRCh38
- UniGene ID
- Hs.131459
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023944.3 |
NP_076433.3
|
10, 11 |
1378 |
|
NR_117085.1 |
|
9, 10 |
1396 |
|
XM_006722850.3 |
XP_006722913.1
|
10, 11 |
1263 |
|
XM_011528202.2 |
XP_011526504.1
|
8, 9 |
898 |
|
XM_011528203.2 |
XP_011526505.1
|
7, 8 |
2120 |
|
XM_011528204.2 |
XP_011526506.1
|
8, 9 |
1599 |
|
XM_011528205.2 |
XP_011526507.1
|
7, 8 |
811 |
|
XM_011528207.2 |
XP_011526509.1
|
7, 8 |
2507 |
|
XM_011528208.2 |
XP_011526510.1
|
9, 10 |
1017 |
|
GenBank mRNA |
AB035130.1 |
- |
10, 11 |
1289 |
|
AB035131.1 |
- |
10, 11 |
1289 |
|
AK074412.1 |
- |
4, 5 |
2680 |
|
AK075435.1 |
- |
9, 10 |
1324 |
|
AL832171.1 |
- |
9, 10 |
2285 |
|
AY008841.1 |
- |
9, 10 |
1259 |
|
AY358977.1 |
- |
10, 11 |
1300 |
|
BC035350.1 |
- |
10, 11 |
1305 |
5187264 |
View Full Product Details