Gene Transcripts
- Gene Symbol
- C10orf76
- Entrez Gene ID
- 79591
- Gene Name
- chromosome 10 open reading frame 76
- Gene Aliases
- Chromosome Location
- Chr. 10: 101845599 - 102056237 on Build GRCh38
- UniGene ID
- Hs.16004
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024541.2 |
NP_078817.2
|
21, 22 |
1682 |
|
XM_011540151.2 |
XP_011538453.1
|
21, 22 |
1727 |
|
XM_011540152.2 |
XP_011538454.1
|
20, 21 |
1614 |
|
XM_011540153.2 |
XP_011538455.1
|
20, 21 |
1578 |
|
XM_011540154.2 |
XP_011538456.1
|
19, 20 |
1512 |
|
XM_011540156.2 |
XP_011538458.1
|
14, 15 |
1646 |
|
XM_011540157.2 |
XP_011538459.1
|
20, 21 |
1631 |
|
XM_017016635.1 |
XP_016872124.1
|
21, 22 |
1680 |
|
XM_017016636.1 |
XP_016872125.1
|
21, 22 |
1680 |
|
GenBank mRNA |
AK023176.1 |
- |
4, 5 |
311 |
|
BC110511.1 |
- |
4, 5 |
311 |
|
View Full Product Details