Gene Transcripts

Gene Symbol
C10orf76
Entrez Gene ID
79591
Gene Name
chromosome 10 open reading frame 76
Gene Aliases
Chromosome Location
Chr. 10: 101845599 - 102056237 on Build GRCh38
UniGene ID
Hs.16004
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024541.2 NP_078817.2 22 1745
XM_011540151.2 XP_011538453.1 22 1790
XM_011540152.2 XP_011538454.1 21 1677
XM_011540153.2 XP_011538455.1 21 1641
XM_011540154.2 XP_011538456.1 20 1575
XM_011540156.2 XP_011538458.1 15 1709
XM_011540157.2 XP_011538459.1 21 1694
XM_017016635.1 XP_016872124.1 22 1743
XM_017016636.1 XP_016872125.1 22 1743
GenBank mRNA AK023176.1 - 5 374
BC110511.1 - 5 374

View Full Product Details