Gene Transcripts

Gene Symbol
EPC1
Entrez Gene ID
80314
Gene Name
enhancer of polycomb homolog 1
Gene Aliases
Epl1
Chromosome Location
Chr. 10: 32267716 - 32378798 on Build GRCh38
UniGene ID
Hs.167805
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001272004.1 NP_001258933.1 1 392
NM_025209.3 NP_079485.1 1 392
NR_104159.1 1 392
NR_104160.1 1 392
GenBank mRNA AF277374.1 - 1 359
AK309822.1 - 1 261
BC036529.1 - 1 393 4828057
BG719217.1 - 1 300
BX476993.1 - 1 383
DA333251.1 - 1 376
DB447845.1 - 1 330

View Full Product Details