Gene Transcripts

Gene Symbol
KAT8
Entrez Gene ID
84148
Gene Name
lysine acetyltransferase 8
Gene Aliases
MOF, MYST1, ZC2HC8, hMOF
Chromosome Location
Chr. 16: 31117664 - 31131393 on Build GRCh38
UniGene ID
Hs.533803
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032188.2 NP_115564.2 5 621
NM_182958.2 NP_892003.2 5 621
XM_011545970.1 XP_011544272.1 6 1162
XM_011545971.1 XP_011544273.1 6 1167
GenBank mRNA AF217501.1 - 5 587
AF260665.2 - 5 519
AF289578.1 - 2 1549
AK021872.1 - 5 618
AK024102.1 - 5 619
AK225660.1 - 5 621
AK291106.1 - 5 618
BC037773.1 - 5 618 5278026

View Full Product Details