Gene Transcripts

Gene Symbol
FAM213A
Entrez Gene ID
84293
Gene Name
family with sequence similarity 213 member A
Gene Aliases
C10orf58, PAMM
Chromosome Location
Chr. 10: 80407829 - 80437115 on Build GRCh38
UniGene ID
Hs.500333
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243778.1 NP_001230707.1 5, 6 647
NM_001243779.1 NP_001230708.1 5, 6 717
NM_001243780.1 NP_001230709.1 5, 6 1213
NM_001243781.1 NP_001230710.1 5, 6 730
NM_001243782.1 NP_001230711.1 5, 6 630
NM_032333.4 NP_115709.3 5, 6 747
XM_011540265.2 XP_011538567.1 5, 6 1050
XM_011540266.2 XP_011538568.1 5, 6 1453
XM_017016775.1 XP_016872264.1 5, 6 734
GenBank mRNA AF305824.1 - 3, 4 403
AK074643.1 - 5, 6 692
AK075447.1 - 5, 6 722
AK225387.1 - 5, 6 621
AK315440.1 - 5, 6 641
AY359059.1 - 5, 6 640
BC005871.2 - 5, 6 609 3010078
BE742158.1 - 5, 6 716
CN396006.1 - 5, 6 599

View Full Product Details