Gene Transcripts
- Gene Symbol
- KIRREL3
- Entrez Gene ID
- 84623
- Gene Name
- kin of IRRE like 3 (Drosophila)
- Gene Aliases
- KIRRE, MRD4, NEPH2, PRO4502
- Chromosome Location
- Chr. 11: 126423493 - 127003460 on Build GRCh38
- UniGene ID
- Hs.376015
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001161707.1 |
NP_001155179.1
|
2, 3 |
490 |
|
NM_001301097.1 |
NP_001288026.1
|
2, 3 |
490 |
|
NM_032531.3 |
NP_115920.1
|
2, 3 |
490 |
|
XM_011543026.2 |
XP_011541328.1
|
2, 3 |
234 |
|
XM_011543027.2 |
XP_011541329.1
|
3, 4 |
695 |
|
XM_011543028.2 |
XP_011541330.1
|
3, 4 |
554 |
|
XM_011543030.2 |
XP_011541332.1
|
2, 3 |
231 |
|
XM_011543031.2 |
XP_011541333.1
|
3, 4 |
689 |
|
XM_011543032.2 |
XP_011541334.1
|
2, 3 |
231 |
|
XM_011543033.1 |
XP_011541335.1
|
3, 4 |
695 |
|
XM_017018418.1 |
XP_016873907.1
|
2, 3 |
785 |
|
XM_017018419.1 |
XP_016873908.1
|
3, 4 |
695 |
|
XM_017018420.1 |
XP_016873909.1
|
3, 4 |
554 |
|
GenBank mRNA |
AB058770.1 |
- |
2, 3 |
379 |
|
AF480410.1 |
- |
2, 3 |
129 |
|
AY358743.1 |
- |
2, 3 |
430 |
|
BC101775.1 |
- |
2, 3 |
215 |
8069281 |
BC101801.1 |
- |
2, 3 |
215 |
8069307 |
View Full Product Details