Gene Transcripts

Gene Symbol
SPATA22
Entrez Gene ID
84690
Gene Name
spermatogenesis associated 22
Gene Aliases
NYD-SP20, NYDSP20
Chromosome Location
Chr. 17: 3440011 - 3513852 on Build GRCh38
UniGene ID
Hs.351068
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170695.1 NP_001164166.1 6 573
NM_001170696.1 NP_001164167.1 5 444
NM_001170697.1 NP_001164168.1 6 579
NM_001170698.1 NP_001164169.1 6 594
NM_001170699.1 NP_001164170.1 6 813
NM_001321336.1 NP_001308265.1 6 863
NM_001321337.1 NP_001308266.1 6 911
NM_032598.4 NP_115987.2 6 635
GenBank mRNA AF367472.1 - 6 527
AK295299.1 - 6 863
AK301892.1 - 6 779
AY032684.1 - 5 393
AY035867.1 - 6 576
AY035868.1 - 6 635
BC029483.1 - 6 520 5271286
BG716998.1 - 6 576

View Full Product Details