Gene Transcripts
- Gene Symbol
- KLHL22
- Entrez Gene ID
- 84861
- Gene Name
- kelch like family member 22
- Gene Aliases
- KELCHL
- Chromosome Location
- Chr. 22: 20441519 - 20496249 on Build GRCh38
- UniGene ID
- Hs.517419
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_032775.3 |
NP_116164.2
|
2, 3 |
375 |
|
XM_017029019.1 |
XP_016884508.1
|
2, 3 |
671 |
|
XM_017029020.1 |
XP_016884509.1
|
3, 4 |
1850 |
|
XM_017029021.1 |
XP_016884510.1
|
2, 3 |
304 |
|
GenBank mRNA |
AK027266.1 |
- |
2, 3 |
314 |
|
AK222850.1 |
- |
2, 3 |
281 |
|
AK290669.1 |
- |
2, 3 |
287 |
|
BC015923.1 |
- |
2, 3 |
327 |
4562420 |
CR456352.1 |
- |
1, 2 |
252 |
|
DA381792.1 |
- |
2, 3 |
375 |
|
View Full Product Details