Gene Transcripts

Gene Symbol
ZSCAN10
Entrez Gene ID
84891
Gene Name
zinc finger and SCAN domain containing 10
Gene Aliases
ZNF206
Chromosome Location
Chr. 16: 3088890 - 3099317 on Build GRCh38
UniGene ID
Hs.334515
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282415.1 NP_001269344.1 5 558
NM_001282416.1 NP_001269345.1 5 753
NM_032805.2 NP_116194.2 6 1021
XM_017023791.1 XP_016879280.1 5 1057
XM_017023792.1 XP_016879281.1 5 1057
GenBank mRNA AK027455.1 - 5 749
AK074736.1 - 5 533
BC111367.1 - 4 1350
BC114452.1 - 4 460

View Full Product Details