Gene Transcripts

Gene Symbol
DISP1
Entrez Gene ID
84976
Gene Name
dispatched RND transporter family member 1
Gene Aliases
DISPA
Chromosome Location
Chr. 1: 222814514 - 223005995 on Build GRCh38
UniGene ID
Hs.528817
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032890.3 NP_116279.2 10 2280
XM_005273335.2 XP_005273392.1 8 2121
XM_006711592.2 XP_006711655.1 9 3068
XM_006711594.3 XP_006711657.1 8 1776
XM_011510072.2 XP_011508374.1 9 2120
XM_011510073.2 XP_011508375.1 9 2598
XM_011510074.2 XP_011508376.1 9 2139
XM_011510075.2 XP_011508377.1 9 2626
XM_011510077.2 XP_011508379.1 8 2062
XM_017002611.1 XP_016858100.1 8 2536
XM_017002612.1 XP_016858101.1 9 1945
XM_017002613.1 XP_016858102.1 7 1905
GenBank mRNA AK026114.1 - 1 170
AK056569.1 - 3 1084
BC011542.1 - 1 415

View Full Product Details