Gene Transcripts

Gene Symbol
ZIC5
Entrez Gene ID
85416
Gene Name
Zic family member 5
Gene Aliases
Chromosome Location
Chr. 13: 99963021 - 99971924 on Build GRCh38
UniGene ID
Hs.508570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033132.3 NP_149123.2 1 1239
GenBank mRNA AF378304.1 - 1 1224

View Full Product Details