Gene Transcripts

Gene Symbol
NAV2
Entrez Gene ID
89797
Gene Name
neuron navigator 2
Gene Aliases
HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2
Chromosome Location
Chr. 11: 19345200 - 20121601 on Build GRCh38
UniGene ID
Hs.502116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111018.1 NP_001104488.1 17 4485
NM_001111019.2 NP_001104489.1 7 1897
NM_001244963.1 NP_001231892.1 18 4878
NM_145117.4 NP_660093.2 17 4809
NM_182964.5 NP_892009.3 17 4809
XM_006718364.3 XP_006718427.1 17 5131
XM_006718365.3 XP_006718428.1 18 5200
XM_006718366.3 XP_006718429.1 18 4364
XM_006718367.3 XP_006718430.1 7 6398
XM_006718368.3 XP_006718431.1 6 6329
XM_006718369.2 XP_006718432.1 7 1893
XM_011520444.2 XP_011518746.1 18 5200
XM_011520445.2 XP_011518747.1 17 5134
XM_011520446.2 XP_011518748.1 18 5200
XM_011520448.2 XP_011518750.1 18 5071
XM_011520449.2 XP_011518751.1 17 5134
XM_011520451.2 XP_011518753.1 17 4578
XM_011520452.2 XP_011518754.1 18 4948
XM_011520453.2 XP_011518755.1 7 1828
XM_017018520.1 XP_016874009.1 19 4992
XM_017018522.1 XP_016874011.1 18 4948
XM_017018523.1 XP_016874012.1 18 5877
XM_017018524.1 XP_016874013.1 18 4700
XM_017018525.1 XP_016874014.1 12 3543
XM_017018526.1 XP_016874015.1 8 4179
GenBank mRNA AB037840.3 - 17 4810
AB063115.1 - 17 4485
AB063116.1 - 17 4778
AF466143.1 - 17 4524
AF466144.1 - 17 4524
AJ488102.1 - 18 4616
AK074287.1 - 7 1897
BC150290.1 - 17 4810 8860130

View Full Product Details