Gene Transcripts

Gene Symbol
ATG16L2
Entrez Gene ID
89849
Gene Name
autophagy related 16 like 2
Gene Aliases
ATG16B, WDR80
Chromosome Location
Chr. 11: 72814406 - 72843740 on Build GRCh38
UniGene ID
Hs.653186
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318766.1 NP_001305695.1 17 1882
NM_033388.1 NP_203746.1 17 1762
XM_005274376.4 XP_005274433.1 17 1761
XM_006718732.2 XP_006718795.1 16 1698
XM_006718734.2 XP_006718797.1 9 2941
XM_011545332.1 XP_011543634.1 16 1647
XM_011545333.1 XP_011543635.1 17 2210
XM_011545334.1 XP_011543636.1 17 1623
GenBank mRNA AK024423.1 - 7 4196
AK090597.1 - 17 1873
AK091151.1 - 16 3350
AK093017.1 - 17 1735
AL832974.1 - 9 1373
BC036713.2 - 15 1491
BC137489.1 - 17 1860 9021489
BC137490.1 - 17 1860 9021490
BC142718.1 - 15 3458
BC146660.1 - 15 3458

View Full Product Details