Gene Transcripts

Gene Symbol
SCRN2
Entrez Gene ID
90507
Gene Name
secernin 2
Gene Aliases
Ses2
Chromosome Location
Chr. 17: 47837681 - 47841333 on Build GRCh38
UniGene ID
Hs.239718
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145023.1 NP_001138495.1 2 161
NM_138355.3 NP_612364.2 2 161
XM_005257776.4 XP_005257833.1 2 141
XM_017025286.1 XP_016880775.1 2 89
GenBank mRNA AK290646.1 - 2 64
AK303835.1 - 2 117
AK308989.1 - 2 89
BC002980.1 - 2 104
BC010408.2 - 2 75 3945715
BC017317.2 - 2 74 3832079

View Full Product Details