Gene Transcripts
- Gene Symbol
- CHMP7
- Entrez Gene ID
- 91782
- Gene Name
- charged multivesicular body protein 7
- Gene Aliases
- Chromosome Location
- Chr. 8: 23243637 - 23262000 on Build GRCh38
- UniGene ID
- Hs.5019
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317899.1 |
NP_001304828.1
|
9, 10 |
1240 |
|
NM_152272.4 |
NP_689485.1
|
10, 11 |
1954 |
|
XM_006716415.2 |
XP_006716478.1
|
8, 9 |
1349 |
|
XM_017013964.1 |
XP_016869453.1
|
10, 11 |
3103 |
|
XM_017013965.1 |
XP_016869454.1
|
8, 9 |
1156 |
|
GenBank mRNA |
AK021776.1 |
- |
9, 10 |
1383 |
|
AK092500.1 |
- |
9, 10 |
1190 |
|
AK097202.1 |
- |
10, 11 |
1951 |
|
AK315664.1 |
- |
9, 10 |
1391 |
|
AL832313.1 |
- |
6, 7 |
1335 |
|
BC004344.1 |
- |
9, 10 |
1182 |
|
BC019110.1 |
- |
9, 10 |
1356 |
5093453 |
BC042050.1 |
- |
10, 11 |
1935 |
5551762 |
View Full Product Details