Gene Transcripts

Gene Symbol
PKD1L2
Entrez Gene ID
114780
Gene Name
polycystin 1 like 2 (gene/pseudogene)
Gene Aliases
PC1L2
Chromosome Location
Chr. 16: 81100879 - 81220370 on Build GRCh38
UniGene ID
Hs.413525
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001076780.1 NP_001070248.1 15 2530
NM_001278423.1 NP_001265352.1 4 624
NM_001278425.1 NP_001265354.1 4 624
NM_052892.3 NP_443124.3 15 2530
NR_126532.1 15 2530
GenBank mRNA AB067466.1 - 7 875
AK098052.1 - 9 1112
AK127266.1 - 3 412
AY164483.1 - 15 2530
AY371495.1 - 4 586
BC004562.2 - 4 591
BC014157.2 - 4 591
CR627341.1 - 2 117

View Full Product Details