Gene Transcripts

Gene Symbol
TRMT61A
Entrez Gene ID
115708
Gene Name
tRNA methyltransferase 61A
Gene Aliases
C14orf172, GCD14, Gcd14p, TRM61, hTRM61
Chromosome Location
Chr. 14: 103529172 - 103537073 on Build GRCh38
UniGene ID
Hs.525610
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152307.2 NP_689520.2 2 379
GenBank mRNA AK097771.1 - 2 379
BC010167.2 - 2 362 3960186
DB496873.1 - 2 379

View Full Product Details