Gene Transcripts

Gene Symbol
SLC16A10
Entrez Gene ID
117247
Gene Name
solute carrier family 16 member 10
Gene Aliases
MCT10, PRO0813, TAT1
Chromosome Location
Chr. 6: 111087503 - 111227125 on Build GRCh38
UniGene ID
Hs.591327
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018593.4 NP_061063.2 3 732
XM_006715329.2 XP_006715392.1 3 807
XM_011535422.2 XP_011533724.1 3 807
XM_017010237.1 XP_016865726.1 2 838
GenBank mRNA AB057445.1 - 3 732
AK126183.1 - 3 807
BC034031.1 - 3 807 4604910
BC066985.1 - 3 704 4827423

View Full Product Details