Gene Transcripts

Gene Symbol
OR52R1
Entrez Gene ID
119695
Gene Name
olfactory receptor family 52 subfamily R member 1 (gene/pseudogene)
Gene Aliases
OR11-22
Chromosome Location
Chr. 11: 4803433 - 4804380 on Build GRCh38
UniGene ID
Hs.553563
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005177.3 NP_001005177.3 1 828

View Full Product Details