Gene Transcripts
- Gene Symbol
- SLC16A14
- Entrez Gene ID
- 151473
- Gene Name
- solute carrier family 16 member 14
- Gene Aliases
- MCT14
- Chromosome Location
- Chr. 2: 230034974 - 230068999 on Build GRCh38
- UniGene ID
- Hs.504317
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152527.4 |
NP_689740.2
|
4, 5 |
1850 |
|
XM_005246353.3 |
XP_005246410.1
|
3, 4 |
1706 |
|
XM_011510751.2 |
XP_011509053.1
|
4, 5 |
1621 |
|
XM_017003480.1 |
XP_016858969.1
|
4, 5 |
1544 |
|
GenBank mRNA |
AK055317.1 |
- |
1, 2 |
300 |
|
AK055356.1 |
- |
2, 3 |
1107 |
|
AK292873.1 |
- |
4, 5 |
1582 |
|
AL833470.1 |
- |
1, 2 |
349 |
|
BC065524.1 |
- |
4, 5 |
1754 |
6058847 |
BX648181.1 |
- |
2, 3 |
1018 |
|
View Full Product Details