Gene Transcripts

Gene Symbol
SLC2A12
Entrez Gene ID
154091
Gene Name
solute carrier family 2 member 12
Gene Aliases
GLUT12, GLUT8
Chromosome Location
Chr. 6: 133987581 - 134052668 on Build GRCh38
UniGene ID
Hs.486508
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145176.2 NP_660159.1 2 1364
XM_006715349.3 XP_006715412.1 2 1381
XM_017010311.1 XP_016865800.1 2 1381
GenBank mRNA AK056554.1 - 1 103
AK122628.1 - 2 1356
AK226098.1 - 2 1235
AL833602.1 - 2 1348
AY046419.1 - 2 1326
BC070149.1 - 2 1298 30371087

View Full Product Details