Gene Transcripts

Gene Symbol
SLC2A12
Entrez Gene ID
154091
Gene Name
solute carrier family 2 member 12
Gene Aliases
GLUT12, GLUT8
Chromosome Location
Chr. 6: 133987581 - 134052668 on Build GRCh38
UniGene ID
Hs.486508
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145176.2 NP_660159.1 2 1472
XM_006715349.3 XP_006715412.1 2 1489
XM_017010311.1 XP_016865800.1 2 1489
GenBank mRNA AK056554.1 - 1 211
AK122628.1 - 2 1464
AK226098.1 - 2 1343
AL833602.1 - 2 1456
AY046419.1 - 2 1434
BC070149.1 - 2 1406 30371087
BX648889.1 - 3 607

View Full Product Details