Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152573.3 | NP_689786.2 | 10 | 1675 | |
XM_005251730.3 | XP_005251787.1 | 9 | 1655 | ||
XM_005251731.3 | XP_005251788.1 | 10 | 2166 | ||
GenBank mRNA | AK290986.1 | - | 10 | 1417 | |
BC023566.1 | - | 10 | 1675 | 4138119 |