Gene Transcripts

Gene Symbol
LRRC55
Entrez Gene ID
219527
Gene Name
leucine rich repeat containing 55
Gene Aliases
Chromosome Location
Chr. 11: 57181747 - 57191714 on Build GRCh38
UniGene ID
Hs.199853
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005210.2 NP_001005210.1 1 264
GenBank mRNA AK127591.1 - 1 264
BC136737.1 - 1 222 9020727
BC136739.1 - 1 222 9020729
BC150572.1 - 1 106

View Full Product Details