Gene Transcripts

Gene Symbol
NUTM1
Entrez Gene ID
256646
Gene Name
NUT midline carcinoma family member 1
Gene Aliases
C15orf55, FAM22H, NUT
Chromosome Location
Chr. 15: 34343315 - 34357737 on Build GRCh38
UniGene ID
Hs.525769
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284292.1 NP_001271221.1 8 2372
NM_001284293.1 NP_001271222.1 7 2278
NM_175741.2 NP_786883.1 8 2457
GenBank mRNA AF482429.1 - 7 2061
AK302656.1 - 8 2372
AK302680.1 - 7 2015
BC114518.1 - 7 2033 40036588
BC114567.1 - 7 2028 40036589

View Full Product Details