Gene Transcripts

Gene Symbol
TINCR
Entrez Gene ID
257000
Gene Name
tissue differentiation-inducing non-protein coding RNA
Gene Aliases
LINC00036, NCRNA00036, PLAC2, onco-lncRNA-16
Chromosome Location
Chr. 19: 5558167 - 5568034 on Build GRCh38
UniGene ID
Hs.515575
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027064.2 3 894
GenBank mRNA BC036545.1 - 3 861
BC120980.1 - 2 507
BC120981.1 - 2 507

View Full Product Details