Gene Transcripts

Gene Symbol
TIPRL
Entrez Gene ID
261726
Gene Name
TOR signaling pathway regulator
Gene Aliases
TIP, TIP41, TIPRL1
Chromosome Location
Chr. 1: 168178845 - 168202114 on Build GRCh38
UniGene ID
Hs.209431
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031800.2 NP_001026970.1 3 561
NM_152902.4 NP_690866.1 3 561
GenBank mRNA AB097034.1 - 3 444
AK313520.1 - 3 561
AL049670.1 - 3 473
BC009506.2 - 3 351 3532344

View Full Product Details