Gene Transcripts

Gene Symbol
PLA2G4D
Entrez Gene ID
283748
Gene Name
phospholipase A2 group IVD
Gene Aliases
cPLA2delta
Chromosome Location
Chr. 15: 42067562 - 42094559 on Build GRCh38
UniGene ID
Hs.380225
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178034.3 NP_828848.3 15, 16 1658
XM_011521467.2 XP_011519769.1 14, 15 1489
GenBank mRNA AB090876.1 - 15, 16 1661
BC034571.1 - 1, 2 95

View Full Product Details