Gene Transcripts

Gene Symbol
OR2L13
Entrez Gene ID
284521
Gene Name
olfactory receptor family 2 subfamily L member 13
Gene Aliases
OR2L14
Chromosome Location
Chr. 1: 247937029 - 248100922 on Build GRCh38
UniGene ID
Hs.372936
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304535.1 NP_001291464.1 2 728
NM_175911.3 NP_787107.1 3 853
XM_011544169.2 XP_011542471.1 3 597
GenBank mRNA BC028158.1 - 3 691

View Full Product Details