Gene Transcripts
- Gene Symbol
- TPRG1
- Entrez Gene ID
- 285386
- Gene Name
- tumor protein p63 regulated 1
- Gene Aliases
- FAM79B
- Chromosome Location
- Chr. 3: 189000774 - 189325304 on Build GRCh38
- UniGene ID
- Hs.729544
- Species
- Homo sapiens
- Gene Symbol
- TPRG1-AS2
- Entrez Gene ID
- 100874027
- Gene Name
- TPRG1 antisense RNA 2
- Gene Aliases
- Chromosome Location
- Chr. 3: 189238686 - 189240594 on Build GRCh38
- UniGene ID
- Hs.616080
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046722.1 |
|
3 |
399 |
|
GenBank mRNA |
BG213396.1 |
- |
3 |
400 |
|
View Full Product Details