Gene Transcripts

Gene Symbol
OR14I1
Entrez Gene ID
401994
Gene Name
olfactory receptor family 14 subfamily I member 1
Gene Aliases
OR5BU1, OR5BU1P
Chromosome Location
Chr. 1: 248681296 - 248685566 on Build GRCh38
UniGene ID
Hs.449973
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004734.1 NP_001004734.1 1 485
XM_005273132.3 XP_005273189.1 2 620
GenBank mRNA BC137188.1 - 1 509
BC137189.1 - 1 509 9021186

View Full Product Details